Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Tuning Solution 2 in dil. Nitric Acid for ICP-MS
VWR® ARISTAR®, Tuning Solution 2 in dil. Nitric Acid for ICP-MS
Catalog # 456542G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Tuning Solution 2 in dil. Nitric Acid for ICP-MS
Catalog # 456542G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Tuning solution 2 in dil. nitric acid ARISTAR® for ICP-MS

Specifications

About this item

13 Components in 2% HNO₃.

13 components 100 mg/L : Ba, Be, Bi, Ce, Cu, Ho, In, Li, Mg, Pb, Tl, U, Y

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.