Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS
VWR®, Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS
Catalog # 456532E
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS
Catalog # 456532E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS

Specifications

About this item

8 Components in 2% HNO₃.

8 Elements 100 mg/L : Ba, Be, Cu, In, Li, Mg, Tl, U

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.