Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Calibration Standard 3 in Hydrochloric Acid 10% for ICP-MS
VWR® ARISTAR®, Multi-Element Calibration Standard 3 in Hydrochloric Acid 10% for ICP-MS
Catalog # 456512A
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Calibration Standard 3 in Hydrochloric Acid 10% for ICP-MS
Catalog # 456512A
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element calibration standard 3 in hydrochloric acid 10% ARISTAR® for ICP-MS

Specifications

About this item

10 Elements in 10% Hydrochloric acid solution.

10 Elements 10 mg/l : Au, Hf, Ir, Pd, Pt, Rh, Ru, Sb, Sn, Te

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.