Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid 5%, ARISTAR® for calibration blank in ICP
Corrosive
Nitric acid 5%, ARISTAR® for calibration blank in ICP
Catalog # 456484N
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Nitric acid 5%, ARISTAR® for calibration blank in ICP
Catalog # 456484N
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349