Order Entry
Switzerland
ContactUsLinkComponent
Phenolphthalein 1% in ethanol 50% pH-indicator
Flammable
Health
Irritant
Phenolphthalein 1% in ethanol 50% pH-indicator
Catalog # 210894R
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Phenolphthalein 1% in ethanol 50% pH-indicator
Catalog # 210894R
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Application test:
    Passes test

Specification Test Results

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913