Order Entry
Switzerland
ContactUsLinkComponent
Aminoallyl-dUTP (AA-dUTP) sodium salt 10 mM in TE buffer
Aminoallyl-dUTP (AA-dUTP) sodium salt 10 mM in TE buffer
Catalog # 40020.
Supplier:  Biotium
CAS Number:  
Aminoallyl-dUTP (AA-dUTP) sodium salt 10 mM in TE buffer
Catalog # 40020.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :589.17 g/mol
Storage Temperature:Freezer
CAS Number:116840-18-7
Formula:C₁₂H₁₇N₃Na₃O₁₄P₃