Order Entry
Switzerland
ContactUsLinkComponent
 
Rat Recombinant IL-2 (Animal free) (from E. coli)
 
undefined
Rat Recombinant IL-2 (Animal free) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: CTLL-2 Proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5,0×10⁵ units/mg
  • Form:
    Lyophilised
  • Gene ID:
    P17108
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Aldesleukin|TCGF|T cell growth factor
  • Protein/peptide name:
    IL-2
  • Purity:
    ≥95%
  • Molecular weight:
    15,6 kDa
  • Sequence:
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together