Guard column cartridges, TCI OPTI-GUARD™-3
Supplier: TCI
Certificates
About this item
The TCI OPTI-GUARD™ series are compact, high-performance, and easy-to-use HPLC guard columns.
- Special fitting style
- Easy hand tight connection
The cartridges need to be mounted in a holder. This series is recommended for purifications using 4,6 mm I.Ø HPLC columns.
Frequently Bought Together