Human Recombinant WISP-2 (Animal Free) (from E. coli)
Nome del fornitore: Shenandoah Biotechnology
Certificati
Informazioni su questo articolo
WNT1-inducible-signaling pathway protein 2 (WISP-2) is a member of the CYR61/CTGF/NOV (CCN) family of regulatory factors. WISP-2 is expressed in ectodermal, mesodermal, and endodermal lineages, including primary osteoblasts, fibroblasts, mesenchymal stem cells, and adipogenic precursor cells. WISP-2 is a canonical WNT ligand that regulates cell proliferation, adhesion, and metastasis. Secreted WISP-2 promotes mesenchymal precursor cell proliferation and maintains them in an undifferentiated state. In bone-forming osteoblasts, WISP-2 promotes osteoblast adhesion and inhibits osteocalcin production.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifiche
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:O76076
- Reconstitution instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Protein/peptide name:WISP-2
- Purity:≥95%
- Molecular weight:24,4 kDa
- Sequence:MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifiche
Frequently Bought Together