Order Entry
Switzerland
ContactUsLinkComponent
Rat Recombinant SHH (Animal free) (from E. coli)
Rat Recombinant SHH (Animal free) (from E. coli)
Certificates
undefined
Rat Recombinant SHH (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Currently undetermined, ED50 ≤Currently undetermined, bioactivity ≥NA
  • Gene ID:
    Q63673
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Shh|HHG-1
  • UniProtKB:
    Q63673
  • Protein/peptide name:
    SHH
  • Purity:
    ≥95%
  • Molecular weight:
    19,9 kDa
  • Sequence:
    MIIGPGRGFG KRQHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKARI HCSVKAENSV AAKSDG
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together