Human Recombinant growth Hormone (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Growth hormone (GH) is an important mitogenic growth factor that is synthesized, stored, and secreted by somatotropic cells of the anterior pituitary gland. GH stimulates growth, cell reproduction, and cell regeneration. In children, GH deficiencies can cause short stature, growth failure, and delayed sexual maturity. Adult GH deficiency presents with reduced lean body mass, increased adiposity, reduced muscle strength, and ultimately premature mortality. GH replacement therapy is used to treat many growth disorders, including Turner syndrome, chronic renal failure, and Prader–Willi syndrome.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Form:Lyophilised
- Gene ID:P01241
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Somatotropin
- Protein/peptide name:Growth Hormone
- Purity:≥95%
- Molecular weight:22,3 kDa
- Sequence:MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF
- Endotoxin level:Low
- Formulation:20 mM sodium bicarbonate, pH 8,0
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together