Order Entry
Switzerland
ContactUsLinkComponent
Mouse Recombinant GM-CSF (from E. coli)
Mouse Recombinant GM-CSF (from E. coli)
Certificates
undefined
Mouse Recombinant GM-CSF (from E. coli)
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: FDCP-1 cell proliferation, ED₅₀ ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
  • Gene ID:
    P01587
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    CSF-2. Pluripoietin-α|mgI-1GM
  • Protein/peptide name:
    GM-CSF (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    14,3 kDa
  • Sequence:
    MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Acetic acid
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together