Human Recombinant IL-6 (Animal free) (from E. coli)
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells. IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130.
- High quality
- Low endotoxin
- Affordable
IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma. IL-6 is often used for growth of hybridoma cell lines. Human IL-6 is active on mouse and rat cells.
These research grade recombinant proteins are typically used as cytokines, growth factors and chemokines in cell based assays and cell culture.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: B9 cell proliferation, ED50 ≤0,025 ng/ml, bioactivity ≥4,0×10⁷ units/mg
- Gene ID:P05231
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IFN-β2|HSF|BSF-2|B-cell differentiation factor|MGI-2
- UniProtKB:P05231
- Protein/peptide name:IL-6
- Purity:≥97%
- Molecular weight:21 kDa
- Sequence:MPVPPGEDSK DVAAPHRQPL TSSERIDKQI RYILDGISAL RKETCNKSNM CESSKEALAE NNLNLPKMAE KDGCFQSGFN EETCLVKIIT GLLEFEVYLE YLQNRFESSE EQARAVQMST KVLIQFLQKK AKNLDAITTP DPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together