DEC-QUAT® 100 Disinfectant
Supplier: VELTEK ASSOCIATES
New Product
Certificates
About this item
DEC-QUAT® 100 is a quaternary ammonium solution for use on hard non-porous environmental surfaces.
- Effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, virucide
- Food and non-food contact sanitizer effective against a broad spectrum of pathogenic bacteria
- Available as both sterile and non-sterile
- Completely traceable and has been validated for sterility and shelf life
This product is designed for use in hospital, medical and health care institutions as a broad-spectrum disinfectant solution on hard, non-porous environmental surfaces such as walls, ceilings, floors, and countertops. Effective against Pseudomonas aeruginosa, Staphylococcus aureus, and Salmonella choleraesuis. Effective against Hepatitis B Virus when contact time is 10 minutes at 1 oz:1 gal of water. Effective against Human Immunodeficiency Virus Type 1 (HIV) after 30 seconds of contact time.
Packaging: Filled sterile by filtration at 0.2 microns. Gamma-irradiated, double-bagged and delivered with lot-specific sterile documentation.
Frequently Bought Together