General-purpose conformable duct tapes, 2903, Scotch®
Supplier: 3M
Certificates
About this item
Scotch® general purpose duct tape 2903 is a versatile, polyethylene film on canvas fabric with a synthetic rubber finish.
- Good initial tack to a wide variety of surfaces
- Highly conformable
- High tensile strength, ideal for bundling
- Tears easily by hand in both directions
- Individually wrapped for easy identification and to keep the roll clean until ready for use
- Tensile strength: 100 [Newton/25 mm]
- PE coated cloth scrim back material
General-purpose, conformable duct tape is ideal for general maintenance, bundling, wrapping, attaching, sealing, and protecting.
Store at 16 to 25 °C and 40 to 65% relative humidity in its original box.