

Dichloromethane, anhydrous ≥99.7% (by GC) stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:14,6063
Boiling Pt:39…40 °C
Melting Pt:–95 °C
Density:1.325
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Packaged under argon.
:Stabilised with amylene
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay (GC):99.7% min
- Water Content (Karl Fischer Titration):0.01% max
- Residue on Evaporation:3 ppm max
Specification Test Results
Frequently Bought Together