ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Capto™ AVB
Affinity chromatography columns, HiTrap™ Capto™ AVB
Supplier:  Cytiva
Certificates
Affinity chromatography columns, HiTrap™ Capto™ AVB
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-3722-12
  • 17-3722-11
  • Max. pressure:
  • 5 bar
  • 5 bar
  • Storage:
  • 4 to 8 °C, 20% ethanol
  • 4 to 8 °C, 20% ethanol