Orders LinkContactUsLinkComponent
Value masking tape 101E
Value masking tape 101E
Supplier:   3M
Value masking tape 101E
Supplier:  3M
Certificates
Value masking tape 101E
Value masking tape 101E
4 Options Available Below
Product Details & Documents
This is a smooth, multi-purpose crepe paper tape for room temperature, indoor, light-duty applications. It is ideal for non-critical paint masking, but also bundling, sealing, attaching, and other applications.
  • Good initial tack to a wide variety of surfaces in different applications
  • Designed for light-duty applications
  • Operating temperature range up to 30 °C
  • Easy to write on for labelling
The cost-effective general-purpose value masking tape is a chamois-colored, 0,12 mm thick adhesive tape with rubber adhesive for general applications (temporary attachment, masking for painting without burning, etc.). It provides conformability on irregular surfaces and around corners, and is hand tearable for easy use.
Product Family Options
Product Information
  • Colour
    Width
    Thickness
    Availability
    Price
  • MMMA101E-18MM
    Masking tape, rubber adhesive, crepe backing
    Masking tape, rubber adhesive, crepe backing
    Beige
    18 mm
    0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 2.10
    SFr. 2.10
    1 * 1 ROU
       SFr. 2.10
  • MMMA101E-24MM
    Masking tape, rubber adhesive, crepe backing
    Masking tape, rubber adhesive, crepe backing
    Beige
    24 mm
    0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 2.80
    SFr. 2.80
    1 * 1 ROU
       SFr. 2.80
  • MMMA101E-30MM
    Masking tape, rubber adhesive, crepe backing
    Masking tape, rubber adhesive, crepe backing
    Beige
    30 mm
    0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 3.40
    SFr. 3.40
    1 * 1 ROU
       SFr. 3.40
  • MMMA101E-48MM
    Masking tape, rubber adhesive, crepe backing
    Masking tape, rubber adhesive, crepe backing
    Beige
    48 mm
    0,12 mm
    Log in for availability
    Log in for availability
    1 * 1 ROU
    1 * 1 ROU - SFr. 5.50
    SFr. 5.50
    1 * 1 ROU
       SFr. 5.50