Non-phthalate PVC vacuum tubing, Nalgene™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene™ lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
- Compare to Tygon® E-3603 tubing
- Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
- Durometer hardness is shore A, 60
- Thick-walled PVC tubing withstands a full vacuum of 760 mm Hg at room temperature
Nalgene™ Non-phthalate PVC vacuum tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening and discolouration from a wide range of laboratory chemicals (demonstrates dimensional stability) and does not impart odour or taste.
Certifications: Meets food additives amendment of the federal food, drug, and cosmetic act. Meets the requirements of the U.S. pharmacopoeia class VI biological tests for plastics, USP <88>. Complies with FDA CFR21 for food packaging, USP VI, cytotoxicity, REACH, latex-free, RoHS, and California proposition 65 standards.
Caution: Not intended for medical use.
Specifications
- Catalog No:
- NALG8702-1210
- NALG8702-0210
- NALG8702-1145
- NALG8702-0145
- NALG8702-1065
- NALG8702-0065
- NALG8702-0055
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement