ContactUsLinkComponent
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Irritant
Health
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Certificates
undefined
Irritant
Health
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Specification Test Results
  • Suitability (for KF-titration):
    test passed
  • Assay of water:
    Max. 0.01 %
  • Number Of Samples:
    Min. 2