ContactUsLinkComponent
Pyridine, anhydrous ≥99.8% for applications requiring low water content, over molecular sieves <0.0050% H₂O, Riedel-de Haën™
undefined
Pyridine, anhydrous ≥99.8% for applications requiring low water content, over molecular sieves <0.0050% H₂O, Riedel-de Haën™
  • Assay (GC):
    Min. 99.8 %
  • Water (Karl Fischer):
    Max. 0.0050 %
Frequently Bought Together