n-Heptane Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65796-200L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay (GC):Min. 99.0 %
- Boiling range:97 - 98 °C
- Density (D 20/4):0.683 - 0.685
- Refractive index (n 20/D):1.3870 - 1.3880
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.0005 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- S-compounds (as S):Max. 0.005 %
- Reaction against H₂SO₄:complying
Specification Test Results
About this item
:
:100.20 g/mol
Boiling Pt:98.2…98.4 °C
Density:0.69 g/cm3
Flash Pt:–4 °C
UN:1206
ADR:3,II
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544