Order Entry
Switzerland
ContactUsLinkComponent
 
Ethanol absolute (taxed), gradient grade for HPLC (for domestic sale in Germany), Burdick & Jackson™
 
Ethanol absolute (taxed), gradient grade for HPLC (for domestic sale in Germany), Burdick & Jackson™

 

  • Assay (GC):
    Min. 99.9 Vol.%
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Acidity:
    Max. 0.0002 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Transmittance at 210 nm:
    Min. 30 %
  • Transmittance at 225 nm:
    Min. 60 %
  • Transmittance at 240 nm:
    Min. 80 %
  • Transmittance from 260 nm:
    Min. 98 %
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb

 

Frequently Bought Together