Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spatula

Spatula

Spatula with spoon and flat end, mirror finish, 225 mm, 50 mm, 38×15 mm
Catalog # HEATHS15909
Supplier:   HEATHROW
Spatula
Catalog # HEATHS15909
Supplier:  HEATHROW
Spatula
Spatula

Spatula with spoon and flat end, mirror finish, 225 mm, 50 mm, 38×15 mm

Price
SFr. 12.00
Log in for availability
1 * 1 ST
Product Details & Documents

This spatula is used to transfer, lift, and handle powders and granular materials.

  • Made from nickel-plated, stainless steel wire
  • Autoclavable

Mirror finish with flat and spoon ends.

Specifications
  • Description:
    Spatula with spoon and flat end, mirror finish
  • Length:
    225 mm
  • Spoon size:
    38×15 mm
  • Blade length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.