Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biohazard Bags

Biohazard Bags

Biohazard disposable bags, Red, PP, 500/case, 203×305 mm, 0,04 mm
Catalog # HEATHS10320
Supplier:   HEATHROW
Biohazard Bags
Catalog # HEATHS10320
Supplier:  HEATHROW
Biohazard Bags
Biohazard Bags

Biohazard disposable bags, Red, PP, 500/case, 203×305 mm, 0,04 mm

Price
256.00 SFr.
Log in for availability
1 * 500 ST
Product Details & Documents

These biohazard waste bags are for personal or general lab use, sizes to suit all needs.

  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 121 °C (250 °F)
  • Disposable

A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French.

Ordering information: Epoxy coated steel wire holder available for bags HEATHS10320.

Documents

Specifications
  • Packed:
    500/case
  • Colour:
    Red
  • Description:
    Biohazard disposable bags
  • L×W:
    203×305 mm
  • Material:
    PP
  • Seq:
    01
  • Thickness:
    0,04 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.