
Tetrabutylammoniumhydroxid 40% in wässriger Lösung, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
UN:3267
ADR:8,II
:GHS05
Frequently Bought Together