Order Entry
Switzerland
ContactUsLinkComponent
Dimethyldichlorosilane solution ~5% in toluene, LiChropur™ for GC derivatisation, Supelco®
Dimethyldichlorosilane solution ~5% in toluene, LiChropur™ for GC derivatisation, Supelco®
Catalog # 08471-500ML
Supplier:  Merck
CAS Number:  
Dimethyldichlorosilane solution ~5% in toluene, LiChropur™ for GC derivatisation, Supelco®
Catalog # 08471-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless to Almost Colorless
  • Appearance (Form):
    Liquid
  • Argentometr. Titration:
    4.5 - 5.5 %
  • Refractive Index N20/D:
    1.491 - 1.495

Specification Test Results

About this item

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane
 :
 :129.06 g/mol
UN:2920
ADR:3(8),I
CAS Number:75-78-5
Formula:C₂H₆Cl₂Si
MDL Number:MFCD00000491