Peqlab HX-1 / HX-2, Blocks for Block Heaters, Digital
Supplier: VWR Collection
Certificates
About this item
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy.
- Versatile range of interchangeable heating blocks work with many size sample tubes and plates
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.
Frequently Bought Together