Order Entry
Switzerland
ContactUsLinkComponent
Titanium(IV) chloride ≥99.5%, GPR RECTAPUR®
Toxic
Corrosive
Titanium(IV) chloride ≥99.5%, GPR RECTAPUR®
Catalog # 87053.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Titanium(IV) chloride ≥99.5%, GPR RECTAPUR®
Catalog # 87053.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99,5 %

Specification Test Results

About this item

 :
 :189.69 g/mol
UN:1838
ADR:6.1 (8),I
 :GHS06
 :GHS05
CAS Number:7550-45-0
Formula:Cl₄Ti
MDL Number:MFCD00011267