ContactUsLinkComponent
Industrial gloves, LIMA, TRIUS®
Industrial gloves, LIMA, TRIUS®
Supplier:  TRIUS SAFETY
Certificates
undefined
Industrial gloves, LIMA, TRIUS®
Supplier:  TRIUS SAFETY
Specifications
  • Catalog No:
  • TRIULIMA/11
  • TRIULIMA/10
  • TRIULIMA/9
  • TRIULIMA/8
  • TRIULIMA/7
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Liner material:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Finish:
  • Palm nitril coating
  • Palm nitril coating
  • Palm nitril coating
  • Palm nitril coating
  • Palm nitril coating
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Cuff length:
  • 7 cm
  • 7 cm
  • 7 cm
  • 7 cm
  • 7 cm
  • Cuff thickness:
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • Finger thickness:
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • Palm thickness:
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • 9 mil
  • EN norm:
  • EN 388:2016 (4123X)
  • EN 388:2016 (4123X)
  • EN 388:2016 (4123X)
  • EN 388:2016 (4123X)
  • EN 388:2016 (4123X)