Order Entry
Switzerland
ContactUsLinkComponent
Urea 99.0-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Urea 99.0-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # HONC15604H-25KG
CAS Number:  
undefined
Urea 99.0-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # HONC15604H-25KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Identity:
    complying
  • Assay:
    99.0 - 100.5 %
  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Melting range:
    132 - 135 °C
  • Insoluble in Ethanol:
    Max. 0.04 %
  • Loss on drying (105°C, 1 h):
    Max. 1.0 %
  • Sulfated ash:
    Max. 0.1 %
  • Ammonium (NH₄):
    Max. 0.05 %
  • Alkalinely reacting substances:
    complying
  • Iron (Fe):
    Max. 0.00005 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Biuret (carbamylurea):
    Max. 0.1 %
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

Specification Test Results

About this item

 :
Melting Pt:133-134 °C
Melting Pt:133…134 °C
Density:1.32 g/cm3 (20 °C)
 :60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022