



n-Heptane ≥98.5% (by GC), Laboratory Reagent
:142-82-5
:
:H₃C(CH₂)₅CH₃
:MFCD00009544
:100.20 g/mol
:13,04679
:–90.5 °C
:0.69 g/cm3 (15 °C)
:–4 °C
:205-563-8
:1206
:3,II
:GHS09
:GHS07
:GHS08
:GHS02
:Ambient
- Refractive Index:n20/D 1.387 (lit.)
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Refractive index (n 20/D):1.385 - 1.389
- Vapor Pressure:40 mmHg ( 20 °C)
- Explosion Limit:7 %
- Vapor Density:3.5 (vs air)
Frequently Bought Together