2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate ≥98.0% (by HPLC), LiChropur™ for chiral derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-isothiocyanatotetrahydro-2H-pyran-3,4,5-triyl triacetate, GITC
CAS Number:14152-97-7
:
Formula:C₁₅H₁₉NO₉S
MDL Number:MFCD00043085
:389.38 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together