Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide ≥98.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 42102-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: (S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)-3-methylbutanamide, FDNP-Val-NH₂
:
:300.2431 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Refrigerator
CAS Number:132679-61-9
Formula:C₁₁H₁₃FN₄O₅
MDL Number:MFCD00155636