Order Entry
Switzerland
ContactUsLinkComponent
Cellulose triacetate, Selectophore™ ionophore, Supelco®
Cellulose triacetate, Selectophore™ ionophore, Supelco®
Catalog # 22199-50G-F
Supplier:  Merck
CAS Number:  
undefined
Cellulose triacetate, Selectophore™ ionophore, Supelco®
Catalog # 22199-50G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :966.845 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:9012-09-3
Formula:C₄₀H₅₄O₂₇