Nitromethane ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:75-52-5
:
Formula:CH₃NO₂
MDL Number:MFCD00007400
Boiling Pt:101.2 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.138 g/cm³ (20 °C)
Molecular Weight:61.04 g/mol
Flash Pt:44 °C
UN:1261
ADR:3 II
Merck Index:13,06644
EINECS:200-876-6
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85984.230 spec_VWRC85984.290 spec_VWRC85984.460
Product specification
Frequently Bought Together