Order Entry
Switzerland
ContactUsLinkComponent
1-Methylimidazole (NMI) ≥99.0% (by GC), GPR RECTAPUR®
Toxic
Corrosive
1-Methylimidazole (NMI) ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Corrosive
1-Methylimidazole (NMI) ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear colourless to yellow liquid
  • Assay (GC):
    min. 99.0 %
  • Density (d20°C/ 4°C) :
    1.034 - 1.036
  • Water:
    max. 0.5 %
  • Identity (IR):
    Passes test

Specification Test Results

Frequently Bought Together