Collagenase Type 2
ABNOP5273
: Abnova
:
- Pk:100 mg
- Storage conditions:Store at 4 °C on dry atmosphere.
- Enzyme name:Collagenase Type 2
Collagenases are enzymes that break the peptide bonds in collagen.
- Lyophilised
- Dialysed
- Contains higher clostripain activity
- Native protein purified from Clostridium histolyticum
- Recommend Usage:Collagenase Assay; Bone, heart, liver, thyroid and salivary primary cell isolation
Activity:>=125 units per mg dry weight. One Unit releases one micromole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5.
Reconstituted in balanced salt solutions (such as Hank's, Earle's and others) at concentrations from 0,05% to 0,5% (w/v). For enzymetic assay: Reconstituted in 0,05 M TES buffer with 0,36 mM calcium chloride, pH 7,5 at a concentration of 1,0 mg/ml.