ContactUsLinkComponent
Peptide and protein MALDI-MS calibration kit, ProteoMass™, Supelco®
Peptide and protein MALDI-MS calibration kit, ProteoMass™, Supelco®
Catalog # MSCAL1-5X1KT
Supplier:  Merck
CAS Number:  
Peptide and protein MALDI-MS calibration kit, ProteoMass™, Supelco®
Catalog # MSCAL1-5X1KT
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    ProteoMass™ Peptide and Protein MALDI-MS Calibration Kit
Specifications
About this item
This kit features a convenient selection of pre-qualified standard peptides, proteins, matrices, and solvents for calibrating and testing matrix assisted laser desorption ionisation (MALDI) mass spectrometers.
Configured for analysing complex mixtures of proteins and peptides (700 to 66,000 Da), MSCAL1 is ideal for evaluating unknown samples or samples containing a broad range of molecular weights. Applications include calibration, tuning, and sensitivity testing of MALDI instruments from all manufacturers.
Delivery: Kit inlcudes 15 components, incuding 10 ProteoMass™ MALDI-MS Standards.