SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Oxalic acid
Oxalic acid 0.1 M (0.2N) eluent concentrate for IC, Supelco®
Oxalic acid 0.1 M (0.2N) eluent concentrate for IC, Supelco®
68487-1L
:
Merck
:
Oxalic acid 0.1 M (0.2N) eluent concentrate for IC, Supelco®
68487-1L
:
Merck
:
Pk:
1 L
Seq:
5037
:
:
90.04 g/mol
:
M-Clarity™ quality level = MQ100
:
144-62-7
:
C₂H₂O₄
:
MFCD00002573