Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium carbonate 0.1 M in water eluent concentrate for IC, Supelco®
Catalog # 56169-1L
Supplier:  Merck
CAS Number:  
undefined
Sodium carbonate 0.1 M in water eluent concentrate for IC, Supelco®
Catalog # 56169-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :105.99 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494