Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydrogen carbonate 1.9-2.1% 0.1 M in water eluent concentrate for IC, Supelco®
Sodium hydrogen carbonate 1.9-2.1% 0.1 M in water eluent concentrate for IC, Supelco®
Catalog # 36486-1L
Supplier:  Merck
CAS Number:  
undefined
Sodium hydrogen carbonate 1.9-2.1% 0.1 M in water eluent concentrate for IC, Supelco®
Catalog # 36486-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :84.01 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528