Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine succinimidyl ester
Cyanine 555NS succinimidyl ester cyanine dye, non-sulphonated
Cyanine 555NS succinimidyl ester cyanine dye, non-sulphonated
Cyanine 555NS succinimidyl ester cyanine dye, non-sulphonated
Catalog # 90118-1MG
Supplier:
Biotium
CAS Number:
Expand image
Cyanine 555NS succinimidyl ester cyanine dye, non-sulphonated
Catalog # 90118-1MG
Supplier:
Biotium
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
Product Details