Glutaraldehyde 25% in aqueous solution for electron microscopy
: Glutaral, Glutardialdehyde, 1,5-Pentanedial
:111-30-8
:
:C₅H₈O₂
:MFCD00007025
:100.12 g/mol
:–6 °C
:Refrigerator
:2922
:8 (6.1),II
:GHS06
:GHS05
:GHS08
:GHS09
- Environmentally Preferable:
- Appearance:clear colorless or yellow liquid
- Assay :23,5 – 26,5%
- Color scale:max. 50 APHA
- pH:min. 3,0
- Density (d20°C/4°C):1,060 - 1,065
- Absorbance A (235 nm; 1:200 diluted with water; 1 cm):max. 0,6
Frequently Bought Together