ContactUsLinkComponent
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Catalog # HICHHI-191
Supplier:  Avantor
undefined
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Catalog # HICHHI-191
Supplier:  Avantor
Specifications
  • Description:
    Standard polymer tubing cutter
  • Pk:
    1
Specifications
About this item
Standard cutter for 1/16" and 1/8" O.D. tubing.
  • Compatible with PEEK and PTFE tubing
  • Flat burr-free cuts
  • Guide holes to ensure precise cutting
Delivery: Includes five replacement blades.