Order Entry
Switzerland
ContactUsLinkComponent
 
Peqlab peqGOLD, RNA Isolation, Total RNA Kit
  13-6834-02
 :  
undefined
Peqlab peqGOLD, RNA Isolation, Total RNA Kit
  13-6834-02
 :  

 

  • Pk:
    200 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    FFPE tissues
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Total RNA kit
  • Seq:
    03
  • Sample size:
    1×10⁷ eukaryotic cells or 10 - 30 mg tissue
  • Description:
    Total RNA Kit

 

 

Easy to use method for isolation up to 100 µg of total RNA from cultured eukaryotic cells and soft tissues <1×10⁷ cells or 30 mg tissue.

  • Fast, 20 minutes or less
  • Safe with no phenol/chloroform extractions
  • Versatile with both spin and vacuum protocols available
  • Homogeniser column reduces RNA column clogging
  • Compatible with peqGOLD DNase I Digestion Kit for lowest level of gDNA co-purification (13-1091-01)
 : See attached manual.