Order Entry
Switzerland
ContactUsLinkComponent
VWR® Peqlab peqGOLD Plasmid Purification, Plasmid Miniprep Kit I
VWR® Peqlab peqGOLD Plasmid Purification, Plasmid Miniprep Kit I
  13-6943-00
 :  
undefined
VWR® Peqlab peqGOLD Plasmid Purification, Plasmid Miniprep Kit I
  13-6943-00
 :  

 

  • Pk:
    5 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Plasmid Miniprep kit I
  • Seq:
    01
  • Sample size:
    1 - 5 ml
  • Description:
    Plasmid Miniprep Kit I

 

 

Plasmid Miniprep Kit I for the easy, quick and efficient extraction of highly pure plasmid DNA. Isolate up to 30 µg of high quality plasmid DNA from 1 to 5 ml bacterial cultures in 30 minutes or less. Plasmid DNA obtained with peqGOLD Plasmid Miniprep kits can directly be used for a wide range of downstream applications.

  • Suitable for the simultaneous processing of multiple samples
  • Short hands-on and preparation times (<15 minutes)
  • No need for organic extractions or precipitations
  • Complete enzymatic degradation of all cellular RNA
  • Efficient removal of enzyme inhibitors and contaminants
 : Each kit includes peqGOLD DNA columns and 2 ml collection tubes, RNase A solution, Solution I, II and III, HBC buffer and DNA wash buffer and elution buffer.