Order Entry
Switzerland
ContactUsLinkComponent
Peqlab peqGOLD, DNA Clean-up, Gel Extraction Kits
Peqlab peqGOLD, DNA Clean-up, Gel Extraction Kits
Supplier:  VWR Chemicals
Certificates
undefined
Peqlab peqGOLD, DNA Clean-up, Gel Extraction Kits
Supplier:  VWR Chemicals
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
  • Sample type:
    Gel matrix
  • Target molecule:
    DNA
  • Sample size:
    Variable
  • Description:
    Gel Extraction Kit

Specifications

Frequently Bought Together