Avantor® Hichrom, Connectors, PEEK Tees and Crosses
Supplier: Avantor
Certificates
About this item
- Use with one PEEK™ or stainless steel tubing
- Maximum operating pressure 3,500psi (241 bar)
Delivery: Includes three or four HI-505 fingertight fittings.
Frequently Bought Together