Order Entry
Switzerland
ContactUsLinkComponent
Ammonium fluoride solution 40% VLSI, Semiconductor grade (Honeywell 17600)
Corrosive
Toxic
Ammonium fluoride solution 40% VLSI, Semiconductor grade (Honeywell 17600)
Catalog # HONC40292H-5L
CAS Number:  
Corrosive
Toxic
Ammonium fluoride solution 40% VLSI, Semiconductor grade (Honeywell 17600)
Catalog # HONC40292H-5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Particles > 0,5 µm:
    Max. 250 P/ml
  • Assay:
    40 - 41 %
  • Residue on ignition (SO₄):
    Max. 10 ppm
  • pH (1 %,20°C):
    6.2 - 7.0
  • Silver (Ag):
    Max. 0.05 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic, antimony (as As):
    Max. 0.03 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.05 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.1 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.05 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Germanium (Ge):
    Max. 0.05 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.05 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.05 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.1 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.05 ppm
  • Silicon (Si):
    Max. 200 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.02 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Ammonium hydrogen difluoride (NH₄HF₂):
    Max. 200 ppm
  • Chloride (Cl):
    Max. 4 ppm
  • Phosphate (PO₄):
    Max. 0.2 ppm
  • Sulfate (SO₄):
    Max. 2 ppm

Specification Test Results

About this item

 :
 :37.04 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.110 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3287
ADR:6.1,III
 :GHS05
 :GHS06
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423